Assay Details
| Gene Symbol: Trappc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003708.1 | NP_001003708.1 | 1 - 2 | 177 | 83 | |
| XM_008766162.1 | XP_008764384.1 | 1 - 2 | 395 | 83 | ||
| GenBank mRNA | AY373378.1 | - | 1 - 2 | 177 | 83 | |
| BC098628.1 | - | 1 - 2 | 184 | 7311789 | 83 | |
| FQ220285.1 | - | 1 - 2 | 200 | 83 | ||
| FQ229310.1 | - | 1 - 2 | 207 | 83 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Trappc4
Genomic Map