Assay Details
| Gene Symbol: Rbm7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108143.2 | NP_001101613.2 | 3 - 4 | 495 | 65 | |
| GenBank mRNA | BC166987.1 | - | 3 - 4 | 495 | 7442233 | 65 |
| FQ229819.1 | - | 402 | 65 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Rbm7
Genomic Map