Assay Details
Gene Symbol: Sdhd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198788.2 | NP_942083.1 | 1 - 2 | 56 | 88 | |
GenBank mRNA | BC059160.1 | - | 1 - 2 | 61 | 6887444 | 88 |
FQ209438.1 | - | 1 - 2 | 57 | 88 | ||
FQ212791.1 | - | 1 - 2 | 59 | 88 | ||
FQ212873.1 | - | 1 - 2 | 59 | 88 | ||
FQ212985.1 | - | 1 - 2 | 58 | 88 | ||
FQ213509.1 | - | 1 - 2 | 58 | 88 | ||
FQ214315.1 | - | 1 - 2 | 86 | 88 | ||
FQ215255.1 | - | 1 - 2 | 63 | 88 | ||
FQ215355.1 | - | 1 - 2 | 57 | 88 | ||
FQ215505.1 | - | 1 - 2 | 58 | 88 | ||
FQ215951.1 | - | 1 - 2 | 58 | 88 | ||
FQ215986.1 | - | 1 - 2 | 57 | 88 | ||
FQ216027.1 | - | 1 - 2 | 58 | 88 | ||
FQ216314.1 | - | 1 - 2 | 64 | 88 | ||
FQ216503.1 | - | 1 - 2 | 61 | 88 | ||
FQ220862.1 | - | 1 - 2 | 57 | 88 | ||
FQ230166.1 | - | 1 - 2 | 50 | 88 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
