Assay Details
| Gene Symbol: Dpagt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199388.1 | NP_955420.1 | 5 - 6 | 863 | 110 | |
| XM_006242921.2 | XP_006242983.1 | 5 - 6 | 1214 | 110 | ||
| GenBank mRNA | BC063184.1 | - | 5 - 6 | 863 | 6887995 | 110 |
| FQ213783.1 | - | 5 - 6 | 940 | 110 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dpagt1
Genomic Map