Assay Details
| Gene Symbol: Neil1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025754.1 | NP_001020925.1 | 3 - 4 | 661 | 85 | |
| XM_006243134.2 | XP_006243196.1 | 3 - 4 | 1475 | 85 | ||
| XM_006243135.2 | XP_006243197.1 | 3 - 4 | 1581 | 85 | ||
| XM_006243136.2 | XP_006243198.1 | 4 - 5 | 657 | 85 | ||
| GenBank mRNA | BC099125.1 | - | 3 - 4 | 661 | 7445605 | 85 |
More Information
Updated Mapping Information
Set Membership: |
|

Neil1
Genomic Map