Assay Details
Gene Symbol: Iqch | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106830.1 | NP_001100300.1 | 6 - 7 | 686 | 63 | |
XM_008766234.1 | XP_008764456.1 | 1 - 2 | 241 | 63 | ||
XM_008766235.1 | XP_008764457.1 | 1 - 2 | 241 | 63 | ||
XM_008766236.1 | XP_008764458.1 | 1 - 2 | 241 | 63 | ||
XM_008766237.1 | XP_008764459.1 | 6 - 7 | 742 | 63 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P476088 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 411743 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 243783 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 414283 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 841695 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 141918 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 592199 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 201805 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 20092 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 259281 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 479497 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 487471 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 818043 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 334771 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 153865 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 799703 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 571233 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 895344 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 349880 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 134180 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 711415 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 505896 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 666485 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 266908 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 795329 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 29304 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 586094 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 189395 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 690296 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 555726 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 248185 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 492425 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 131297 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 44939 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 244056 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 444110 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 552861 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 43347 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 967026 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 650361 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 543281 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 535114 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 724424 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 174535 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 215540 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 105433 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 373315 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 270178 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 351966 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 203008 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 873518 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 593751 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 489020 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 24301 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 934962 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 628112 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 960787 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 176172 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 207201 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 718634 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 714559 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 45149 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 619033 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 702766 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 967798 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 667056 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 226211 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 929281 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 469806 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 382966 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 708033 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 281992 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 79223 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 935517 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 380433 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 615972 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 579490 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 785472 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 726788 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 107439 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 953595 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 322275 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 775192 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 65199 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 161710 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 844370 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 348982 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 698593 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 319056 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 309475 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 329403 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 230950 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 556857 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 244890 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 7891 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 256787 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 353615 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 85087 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 549976 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 146279 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 461168 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 832736 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 514442 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 26865 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 852836 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 334552 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 666278 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 249603 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 235190 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 646962 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 8907 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 383030 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 525975 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 927048 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 496044 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 365388 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 524468 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 31048 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 122186 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 725905 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 883614 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 356647 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 345784 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 607004 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 953507 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 652337 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 428688 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 643098 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 525635 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 715039 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 390075 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 213348 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 957355 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 535722 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 277495 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 761261 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 957159 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 58068 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 30789 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 89392 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 161748 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 36715 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 39691 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 115195 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 107901 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 87911 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 18122 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 92506 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 94125 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 173349 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 148123 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 172759 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 75879 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 66645 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 165060 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 82383 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 146656 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 26850 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 140367 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 49915 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 114441 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P476088 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P476088 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P476088 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P476088 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 27416 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 68125 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 42877 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 49391 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10918167 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10918166 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10918154 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10918165 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10918150 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10918152 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10918164 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10918168 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10918163 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10918158 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10918162 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10918160 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10918153 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10918157 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10918155 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10918161 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10918159 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10918148 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10918151 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10918149 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10918156 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10918147 | - | - |
Affymetrix Rat Clariom S Assay | PSR0800022226.rn.1 | TC0800002734.rn.1 | RTA TC |
Affymetrix Rat Gene 1.0 ST Array | 10918147 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21237008 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1373631 | - | - |
