Assay Details
| Gene Symbol: Rbpms2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001173426.1 | NP_001166897.1 | 7 - 8 | 716 | 123 | |
| XM_006243274.2 | XP_006243336.1 | 7 - 8 | 2402 | 123 | ||
| XM_008766329.1 | XP_008764551.1 | 6 - 7 | 825 | 123 | ||
| GenBank mRNA | BC169038.1 | - | 7 - 8 | 716 | 9090232 | 123 |
More Information
Updated Mapping Information
Set Membership: |
|

Rbpms2
Genomic Map