Assay Details
| Gene Symbol: Ptpn9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013040.1 | NP_001013058.1 | 4 - 5 | 442 | 68 | |
| XM_006243105.2 | XP_006243167.1 | 4 - 5 | 1019 | 68 | ||
| XM_006243106.2 | XP_006243168.1 | 4 - 5 | 1034 | 68 | ||
| GenBank mRNA | BC082041.1 | - | 4 - 5 | 442 | 7131532 | 68 |
More Information
Updated Mapping Information
Set Membership: |
|

Ptpn9
Genomic Map