Assay Details
Gene Symbol: Hexa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004443.1 | NP_001004443.1 | 5 - 6 | 603 | 88 | |
XM_006243162.1 | XP_006243224.1 | 6 - 7 | 473 | 88 | ||
GenBank mRNA | BC082097.1 | - | 5 - 6 | 603 | 7134440 | 88 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
