Assay Details
| Gene Symbol: Tmem189 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113752.1 | NP_001107224.1 | 2 - 3 | 352 | 94 | |
| GenBank mRNA | BC158660.1 | - | 2 - 3 | 352 | 8369614 | 94 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Tmem189
Genomic Map