Assay Details
| Gene Symbol: Rims4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_170666.1 | NP_733766.1 | 2 - 3 | 242 | 73 | |
| XM_008762471.1 | XP_008760693.1 | 2 - 3 | 359 | 73 | ||
| GenBank mRNA | AF548739.1 | - | 2 - 3 | 242 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P035037 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 796504 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 126725 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 618454 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 122047 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 847073 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 91680 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 747210 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 458098 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 556643 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 617323 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 606334 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 854595 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 1254 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 226888 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 953472 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 785554 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 95124 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 970824 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 63695 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 448008 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 333500 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 715240 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 229578 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 800451 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 712799 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 751235 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 967609 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 499244 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 588284 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 62959 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 251170 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 29607 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 168334 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 780135 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 180219 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 254759 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 291528 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 799471 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 488338 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 563172 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 23335 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 818508 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 744572 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 775868 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 882972 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 898911 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 858033 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 34750 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 8018 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 145958 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 56083 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 76429 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 66170 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 149944 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P035037 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P035037 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P035037 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P259727 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 13771 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 58280 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 36726 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 45240 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 58740 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10851533 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10851528 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10851529 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10851530 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10851532 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10851531 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10851527 | - | - |
| Affymetrix Rat Clariom S Assay | PSR0300036155.rn.1 | TC0300004723.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0300036152.rn.1 | TC0300004723.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0300036157.rn.1 | TC0300004723.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0300036153.rn.1 | TC0300004723.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0300036156.rn.1 | TC0300004723.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0300036154.rn.1 | TC0300004723.rn.1 | RTA TC |
| Affymetrix Rat Gene 1.0 ST Array | 10851527 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1350612 | - | - |
| RatRef-12 Expression BeadChip | ILMN_63783 | - | - |

Rims4
Genomic Map