Assay Details
| Gene Symbol: Cd40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134360.1 | NP_599187.1 | 6 - 7 | 582 | 105 | |
| XM_006235511.2 | XP_006235573.1 | 6 - 7 | 644 | 105 | ||
| XM_008762464.1 | XP_008760686.1 | 6 - 7 | 643 | 105 | ||
| GenBank mRNA | BC097949.1 | - | 6 - 7 | 582 | 7387440 | 105 |
| FQ226992.1 | - | 6 - 7 | 565 | 105 | ||
| FQ231420.1 | - | 6 - 7 | 585 | 105 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cd40
Genomic Map