Assay Details
Gene Symbol: LOC499179 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008759598.1 | XP_008757820.1 | 4 | 1963 | 81 |
Gene Symbol: Top1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022615.1 | NP_072137.1 | 17 - 18 | 1831 | 81 | |
GenBank mRNA | AF140782.1 | - | 17 - 18 | 1831 | 81 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
DNA replication
DNA topological change
chromatin remodeling
chromosome segregation
circadian rhythm
response to temperature stimulus
rRNA transcription
response to gamma radiation
organ regeneration
circadian regulation of gene expression
embryonic cleavage
response to drug
response to cAMP
cellular response to luteinizing hormone stimulus
core promoter sequence-specific DNA binding
DNA binding
chromatin binding
DNA topoisomerase type I activity
DNA topoisomerase type II (ATP-hydrolyzing) activity
chromatin DNA binding
protein complex binding
poly(A) RNA binding
DNA replication
DNA topological change
chromatin remodeling
chromosome segregation
circadian rhythm
response to temperature stimulus
rRNA transcription
response to gamma radiation
organ regeneration
circadian regulation of gene expression
embryonic cleavage
response to drug
response to cAMP
cellular response to luteinizing hormone stimulus
core promoter sequence-specific DNA binding
DNA binding
chromatin binding
DNA topoisomerase type I activity
DNA topoisomerase type II (ATP-hydrolyzing) activity
chromatin DNA binding
protein complex binding
poly(A) RNA binding
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P241862 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P031456 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 620045 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 253219 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 634974 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 356591 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 498197 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 825717 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 244608 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 854276 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 40245 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 626751 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 692854 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 258502 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 837480 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 561341 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 408155 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 39 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 160652 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 811505 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 823580 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 130595 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 449458 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 228823 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 416244 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 242332 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 845159 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 260639 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 681268 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 503339 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 247297 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 107937 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 230317 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 176527 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 416746 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 638322 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 451211 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 97908 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 138893 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 122885 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 667510 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 584017 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 939707 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 617599 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 913219 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 619893 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 914683 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 926522 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 163131 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 598650 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 316517 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 45640 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 809667 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 341454 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 308794 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 451610 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 555797 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 72837 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 758709 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 399370 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 349786 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 815620 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 291766 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 695056 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 46513 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 70504 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 21455 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 90225 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 8036 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 79448 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 169081 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 80959 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 29228 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 13678 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 176311 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 39313 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P031456 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P241862 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P241862 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P031456 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P241862 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P031456 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P438054 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P241862 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 53625 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 31135 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 24199 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 84020 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 76844 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 7830 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10841835 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10841838 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10841840 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10707940 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10841830 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10841849 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10841843 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10841846 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10841842 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10841839 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10841841 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10841831 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10841845 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10841832 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10841833 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10841834 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10841848 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10841847 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10841836 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10841844 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10841837 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10707941 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10707939 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10841829 | - | - |
Rat Genome 230 2.0 Array | 1369421_at | - | - |
Rat Genome 230 2.0 Array | 1399046_at | - | - |
Rat Genome 230 2.0 Array | 1369421_PM_at | - | - |
Rat Genome 230 2.0 Array | 1399046_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1369421_at | - | - |
Affymetrix HT Rat Focus Array | 1369421_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0300016468.rn.1 | TC0300002215.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0300016471.rn.1 | TC0300002215.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0300016469.rn.1 | TC0300002215.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0300016470.rn.1 | TC0300002215.rn.1 | RTA TC |
Rat Expression Set 230 | 1399046_PM_at | - | - |
Rat Expression Set 230 | 1369421_PM_at | - | - |
Rat Expression Set 230 | 1399046_at | - | - |
Rat Expression Set 230 | 1369421_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10841829 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10707939 | - | - |
Rat Genome U34 Set | AA848218_at | - | - |
Rat Genome U34 Set | rc_AA819468_at | - | - |
Rat Genome U34 Set | AA848218_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1349123 | - | - |
RatRef-12 Expression BeadChip | ILMN_56331 | - | - |
