Assay Details
| Gene Symbol: Ptpn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012637.2 | NP_036769.1 | 5 - 6 | 566 | 71 | |
| XM_006235638.2 | XP_006235700.1 | 5 - 6 | 635 | 71 | ||
| XM_006235639.2 | XP_006235701.1 | 5 - 6 | 636 | 71 | ||
| GenBank mRNA | BC081829.1 | - | 5 - 6 | 566 | 7135855 | 71 |
| M33962.1 | - | 5 - 6 | 608 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
protein dephosphorylation
insulin receptor signaling pathway
regulation of signal transduction
regulation of endocytosis
negative regulation of vascular endothelial growth factor receptor signaling pathway
endoplasmic reticulum unfolded protein response
actin cytoskeleton reorganization
peptidyl-tyrosine dephosphorylation
platelet-derived growth factor receptor-beta signaling pathway
regulation of insulin receptor signaling pathway
negative regulation of ERK1 and ERK2 cascade
regulation of hepatocyte growth factor receptor signaling pathway
peptidyl-tyrosine dephosphorylation involved in inactivation of protein kinase activity
protein tyrosine phosphatase activity
insulin receptor binding
protein binding
zinc ion binding
enzyme binding
protein kinase binding
receptor tyrosine kinase binding
poly(A) RNA binding
ephrin receptor binding
protein phosphatase 2A binding
protein dephosphorylation
insulin receptor signaling pathway
regulation of signal transduction
regulation of endocytosis
negative regulation of vascular endothelial growth factor receptor signaling pathway
endoplasmic reticulum unfolded protein response
actin cytoskeleton reorganization
peptidyl-tyrosine dephosphorylation
platelet-derived growth factor receptor-beta signaling pathway
regulation of insulin receptor signaling pathway
negative regulation of ERK1 and ERK2 cascade
regulation of hepatocyte growth factor receptor signaling pathway
peptidyl-tyrosine dephosphorylation involved in inactivation of protein kinase activity
protein tyrosine phosphatase activity
insulin receptor binding
protein binding
zinc ion binding
enzyme binding
protein kinase binding
receptor tyrosine kinase binding
poly(A) RNA binding
ephrin receptor binding
protein phosphatase 2A binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P132432 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P132437 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 433536 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 724606 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 854083 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 500163 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 338874 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 255354 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 263530 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 227306 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 720293 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 673813 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 670402 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 167605 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 238610 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 704415 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 573450 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 232573 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 244493 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 450737 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 25787 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 268179 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 491277 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 217391 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 544320 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 814402 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 588409 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 121494 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 714101 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 519591 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 424831 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 46060 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 863234 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 591622 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 321928 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 301543 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 105195 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 138222 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 97352 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 117891 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 42449 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P132432 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P132437 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P132437 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P132432 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P132437 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P132432 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P519834 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P11497 | - | - |
| Matsumoto (Agilent Rat cDNA Microarray G4105A) | 700493485 | - | - |
| Matsumoto (Agilent Rat cDNA Microarray G4105A) | 700513145 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 63770 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 44484 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 96626 | - | - |
| Agilent Rat cDNA Microarray (G4105A) | 700493485 | - | - |
| Agilent Rat cDNA Microarray (G4105A) | 700513145 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10842476 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10842482 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10842484 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10842480 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10842483 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10842479 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10842485 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10842481 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10842477 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10842478 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10842475 | - | - |
| Rat Genome 230 2.0 Array | 1397916_s_at | - | - |
| Rat Genome 230 2.0 Array | 1368645_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1397916_PM_s_at | - | - |
| Rat Genome 230 2.0 Array | 1368645_at | - | - |
| Affymetrix HT Rat Focus Array | 1368645_at | - | - |
| Affymetrix HT Rat Focus Array | 1368645_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1397916_PM_s_at | - | - |
| Affymetrix HT Rat Focus Array | 1397916_s_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0300017799.rn.1 | TC0300002381.rn.1 | RTA TC |
| Rat Expression Set 230 | 1368645_PM_at | - | - |
| Rat Expression Set 230 | 1368645_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10842475 | - | - |
| Rat Genome U34 Set | M33962_at | - | - |
| Rat Genome U34 Set | M33962_g_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 22345955 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1355065 | - | - |
| RatRef-12 Expression BeadChip | ILMN_70340 | - | - |

Ptpn1
Genomic Map