Assay Details
| Gene Symbol: Tnnc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037351.1 | NP_001032428.1 | 1 - 2 | 54 | 86 | |
| GenBank mRNA | DQ273679.1 | - | 1 - 2 | 54 | 86 | |
| FQ215069.1 | - | 118 | 86 | |||
| FQ216531.1 | - | 118 | 86 | |||
| FQ216869.1 | - | 118 | 86 | |||
| FQ216875.1 | - | 148 | 86 | |||
| FQ216940.1 | - | 110 | 86 | |||
| FQ216956.1 | - | 131 | 86 | |||
| FQ217047.1 | - | 118 | 86 | |||
| FQ217054.1 | - | 118 | 86 | |||
| FQ217106.1 | - | 147 | 86 | |||
| FQ217137.1 | - | 118 | 86 | |||
| FQ217169.1 | - | 118 | 86 | |||
| FQ217172.1 | - | 118 | 86 | |||
| FQ217195.1 | - | 118 | 86 | |||
| FQ217236.1 | - | 129 | 86 | |||
| FQ217299.1 | - | 118 | 86 | |||
| FQ217371.1 | - | 118 | 86 | |||
| FQ217378.1 | - | 118 | 86 | |||
| FQ217706.1 | - | 118 | 86 | |||
| FQ217788.1 | - | 118 | 86 | |||
| FQ217840.1 | - | 118 | 86 | |||
| FQ217842.1 | - | 118 | 86 | |||
| FQ217896.1 | - | 117 | 86 | |||
| FQ217902.1 | - | 118 | 86 | |||
| FQ217974.1 | - | 147 | 86 | |||
| FQ217984.1 | - | 118 | 86 | |||
| FQ218010.1 | - | 118 | 86 | |||
| FQ218048.1 | - | 148 | 86 | |||
| FQ218061.1 | - | 118 | 86 | |||
| FQ223543.1 | - | 118 | 86 | |||
| FQ223572.1 | - | 118 | 86 | |||
| FQ223583.1 | - | 118 | 86 | |||
| FQ223607.1 | - | 118 | 86 | |||
| FQ223623.1 | - | 118 | 86 | |||
| FQ223636.1 | - | 118 | 86 | |||
| FQ223643.1 | - | 118 | 86 | |||
| FQ223690.1 | - | 118 | 86 | |||
| FQ223692.1 | - | 118 | 86 | |||
| FQ223711.1 | - | 118 | 86 | |||
| FQ223713.1 | - | 118 | 86 | |||
| FQ223719.1 | - | 118 | 86 | |||
| FQ223773.1 | - | 130 | 86 | |||
| FQ223816.1 | - | 139 | 86 | |||
| FQ223825.1 | - | 118 | 86 | |||
| FQ223832.1 | - | 118 | 86 | |||
| FQ223835.1 | - | 148 | 86 | |||
| FQ223976.1 | - | 118 | 86 | |||
| FQ223986.1 | - | 118 | 86 | |||
| FQ224079.1 | - | 118 | 86 | |||
| FQ224114.1 | - | 147 | 86 | |||
| FQ224223.1 | - | 118 | 86 | |||
| FQ224258.1 | - | 118 | 86 | |||
| FQ224340.1 | - | 118 | 86 | |||
| FQ224376.1 | - | 133 | 86 | |||
| FQ224387.1 | - | 118 | 86 | |||
| FQ224450.1 | - | 118 | 86 | |||
| FQ224462.1 | - | 118 | 86 | |||
| FQ224599.1 | - | 118 | 86 | |||
| FQ224648.1 | - | 118 | 86 | |||
| FQ224672.1 | - | 118 | 86 | |||
| FQ224735.1 | - | 119 | 86 | |||
| FQ224749.1 | - | 118 | 86 | |||
| FQ224755.1 | - | 147 | 86 | |||
| FQ224795.1 | - | 118 | 86 | |||
| FQ224811.1 | - | 118 | 86 | |||
| FQ224822.1 | - | 118 | 86 | |||
| FQ224827.1 | - | 117 | 86 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Tnnc2
Genomic Map