Assay Details
Gene Symbol: Sts | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012661.1 | NP_036793.1 | 4 - 5 | 922 | 61 | |
XM_006256960.2 | XP_006257022.1 | 4 - 5 | 1169 | 61 | ||
XM_006256961.2 | XP_006257023.1 | 4 - 5 | 1157 | 61 | ||
GenBank mRNA | U37138.1 | - | 4 - 5 | 922 | 61 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
