Assay Details
Gene Symbol: Ppef1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034935.1 | NP_001030107.1 | 14 - 15 | 1866 | 161 | |
GenBank mRNA | BC104696.1 | - | 14 - 15 | 1866 | 7112183 | 161 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P419710 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 648067 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 913132 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 700638 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 120194 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 438010 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 195554 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 332379 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 665027 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 88962 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 728223 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 663589 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 166954 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 250335 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 744655 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 777765 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 471653 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 930010 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 702103 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 246258 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 636166 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 158839 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 182600 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 609706 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 179782 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 747534 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 462663 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 821211 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 311745 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 401432 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 331125 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 783355 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 704138 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 338115 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 647188 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 12288 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 223636 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 300992 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 133521 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 380626 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 275105 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 705524 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 765282 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 733143 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 802200 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 947132 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 321566 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 351708 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 564828 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 83363 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 169255 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 49868 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 14803 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 170648 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 163915 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 124682 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 51650 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P419710 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P419710 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P419710 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P419710 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 40655 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 2166 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 50444 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 2362 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 1198 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 73484 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 15773 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 65327 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10933564 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10933572 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10933567 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10933573 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10933563 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10933570 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10933560 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10933565 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10933561 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10933571 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10933569 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10933562 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10933568 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10933566 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10933559 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10933559 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21711474 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1360777 | - | - |
