Assay Details
Gene Symbol: Apoo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199178.1 | NP_001186107.1 | 2 - 3 | 123 | 92 | |
XM_008773262.1 | XP_008771484.1 | 2 - 3 | 823 | 92 | ||
GenBank mRNA | FM122174.1 | - | 2 - 3 | 123 | 92 | |
FQ210589.1 | - | 2 - 3 | 165 | 92 | ||
FQ218438.1 | - | 2 - 3 | 138 | 92 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008773262.1 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
