Assay Details
| Gene Symbol: Rpl39l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195471.1 | NP_001182400.1 | 2 - 3 | 133 | 86 | |
| GenBank mRNA | BC103490.1 | - | 2 - 3 | 133 | 86 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC103490.1,NM_001195471.1 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Rpl39l
Genomic Map