Assay Details
| Gene Symbol: Glyr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007800.1 | NP_001007801.1 | 2 - 3 | 77 | 74 | |
| XM_006245803.2 | XP_006245865.1 | 2 - 3 | 146 | 74 | ||
| XM_006245804.2 | XP_006245866.1 | 2 - 3 | 146 | 74 | ||
| GenBank mRNA | BC085931.1 | - | 2 - 3 | 88 | 7111787 | 74 |
More Information
Updated Mapping Information
Set Membership: |
|

Glyr1
Genomic Map