Assay Details
| Gene Symbol: LOC100362572 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008767466.1 | XP_008765688.1 | 1 - 2 | 383 | 83 | |
| XM_008767467.1 | XP_008765689.1 | 1 - 2 | 398 | 83 | ||
More Information
Set Membership: |
|

LOC100362572
Genomic Map