Assay Details
| Gene Symbol: LOC103689979 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006245705.2 | XP_006245767.1 | 1 - 2 | 129 | 89 | |
| XM_006245707.2 | XP_006245769.1 | 1 - 2 | 129 | 89 | ||
| XM_008767471.1 | XP_008765693.1 | 1 - 2 | 189 | 89 | ||
| Gene Symbol: Pdxdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134961.1 | NP_001128433.1 | 195 | 89 | ||
| GenBank mRNA | BC169099.1 | - | 98 | 5623508 | 89 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC169099.1,NM_001134961.1,XM_006245705.2,XM_006245707.2,XM_008767471.1 transcripts.
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

LOC103689979
Genomic Map