Assay Details
| Gene Symbol: LOC103689979 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006245705.2 | XP_006245767.1 | 21 - 22 | 2097 | 131 | |
| XM_006245707.2 | XP_006245769.1 | 20 - 21 | 2034 | 131 | ||
| XM_006245708.2 | XP_006245770.1 | 21 - 22 | 2114 | 131 | ||
| XM_008767471.1 | XP_008765693.1 | 21 - 22 | 2160 | 131 | ||
| Gene Symbol: Pdxdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134961.1 | NP_001128433.1 | 2166 | 131 | ||
| GenBank mRNA | BC169099.1 | - | 2069 | 5623508 | 131 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

LOC103689979
Genomic Map