Assay Details
| Gene Symbol: LOC103689979 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006245705.2 | XP_006245767.1 | 9 - 10 | 918 | 83 | |
| XM_006245707.2 | XP_006245769.1 | 8 - 9 | 855 | 83 | ||
| XM_006245708.2 | XP_006245770.1 | 9 - 10 | 935 | 83 | ||
| XM_008767471.1 | XP_008765693.1 | 9 - 10 | 981 | 83 | ||
| Gene Symbol: Pdxdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134961.1 | NP_001128433.1 | 987 | 83 | ||
| GenBank mRNA | BC169099.1 | - | 890 | 5623508 | 83 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

LOC103689979
Genomic Map