Assay Details
Gene Symbol: Sec14l5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001135710.1 | NP_001129182.1 | 2 - 3 | 207 | 100 | |
XM_006245783.2 | XP_006245845.1 | 2 - 3 | 253 | 100 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P097715 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 822268 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 100317 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 450614 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 824659 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 280601 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 742592 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 555896 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 244561 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 122496 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 389756 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 285189 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 228088 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 458899 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 48216 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 225837 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 495233 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 647037 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 749070 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 950399 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 371506 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 711127 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 368956 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 469679 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 78945 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 274992 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 692992 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 828649 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 972401 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 38326 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 97707 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 137467 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 80547 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P097715 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P097715 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P097715 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P555850 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P113035 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10740614 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10740611 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10740615 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10740613 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10740600 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10740601 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10740604 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10740606 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10740610 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10740605 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10740609 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10740602 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10740607 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10740603 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10740608 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10740612 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10740599 | - | - |
Affymetrix Rat Clariom S Assay | PSR1000020280.rn.1 | TC1000002290.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1000020276.rn.1 | TC1000002290.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1000020279.rn.1 | TC1000002290.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1000020289.rn.1 | TC1000002290.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1000020278.rn.1 | TC1000002290.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1000020282.rn.1 | TC1000002290.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1000020284.rn.1 | TC1000002290.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1000020277.rn.1 | TC1000002290.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1000020288.rn.1 | TC1000002290.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1000020290.rn.1 | TC1000002290.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1000020283.rn.1 | TC1000002290.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1000020281.rn.1 | TC1000002290.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1000020287.rn.1 | TC1000002290.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1000020285.rn.1 | TC1000002290.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1000020286.rn.1 | TC1000002290.rn.1 | RTA TC |
Affymetrix Rat Gene 1.0 ST Array | 10740599 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21043399 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1359511 | - | - |
