Assay Details
| Gene Symbol: Il12rb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191750.1 | NP_001178679.1 | 14 - 15 | 2096 | 79 | |
| XM_006236613.2 | XP_006236675.1 | 16 - 17 | 2490 | 79 | ||
| XM_006236614.2 | XP_006236676.1 | 16 - 17 | 2486 | 79 | ||
| XM_008762975.1 | XP_008761197.1 | 15 - 16 | 2407 | 79 | ||
| GenBank mRNA | AF083329.1 | - | 3 - 4 | 250 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Il12rb2
Genomic Map