Assay Details
| Gene Symbol: Mrpl19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029898.1 | NP_001025069.1 | 1 - 2 | 230 | 111 | |
| GenBank mRNA | FQ235078.1 | - | 1 - 2 | 248 | 111 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mrpl19
Genomic Map