Assay Details
Gene Symbol: Gkn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198972.1 | NP_945323.1 | 1 - 2 | 53 | 55 | |
GenBank mRNA | AY456958.1 | - | 1 - 2 | 53 | 55 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AY456958.1, NM_198972.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
