Assay Details
| Gene Symbol: Gkn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198972.1 | NP_945323.1 | 4 - 5 | 352 | 73 | |
| GenBank mRNA | AY456958.1 | - | 4 - 5 | 352 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gkn1
Genomic Map