Assay Details
| Gene Symbol: Tex261 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017537.1 | NP_001017537.1 | 1 - 2 | 94 | 64 | |
| XM_008763059.1 | XP_008761281.1 | 1 - 2 | 172 | 64 | ||
| GenBank mRNA | BC091302.1 | - | 1 - 2 | 94 | 7309682 | 64 |
| FQ214220.1 | - | 1 - 2 | 106 | 64 | ||
| FQ218231.1 | - | 1 - 2 | 105 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tex261
Genomic Map