Assay Details
Gene Symbol: Prickle2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107876.1 | NP_001101346.1 | 3 - 4 | 380 | 100 | |
XM_006236912.2 | XP_006236974.1 | 3 - 4 | 1019 | 100 | ||
XM_006236913.2 | XP_006236975.1 | 3 - 4 | 5329 | 100 | ||
XM_008763124.1 | XP_008761346.1 | 3 - 4 | 1020 | 100 | ||
XM_008763125.1 | XP_008761347.1 | 3 - 4 | 358 | 100 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
