Assay Details
Gene Symbol: Ccdc37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006236885.2 | XP_006236947.1 | 15 - 16 | 2436 | 90 | |
XM_006236886.2 | XP_006236948.1 | 14 - 15 | 2260 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() |
