Assay Details
| Gene Symbol: Eif4e3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106612.1 | NP_001100082.1 | 2 - 3 | 216 | 78 | |
| XM_008763159.1 | XP_008761381.1 | 2 - 3 | 630 | 78 | ||
| XM_008763160.1 | XP_008761382.1 | 4 - 5 | 700 | 78 | ||
| XM_008763161.1 | XP_008761383.1 | 3 - 4 | 564 | 78 | ||
| GenBank mRNA | BC168936.1 | - | 2 - 3 | 203 | 7124999 | 78 |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008763160.1, XM_008763161.1 transcripts.
Set Membership: |
|

Eif4e3
Genomic Map