Assay Details
| Gene Symbol: Nfu1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106606.2 | NP_001100076.2 | 2 - 3 | 162 | 137 | |
| XM_008763063.1 | XP_008761285.1 | 2 - 3 | 395 | 137 | ||
| GenBank mRNA | FM058785.1 | - | 2 - 3 | 162 | 137 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008763063.1 transcripts.
Set Membership: |
|

Nfu1
Genomic Map