Assay Details
| Gene Symbol: Cdh19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009448.1 | NP_001009448.1 | 8 - 9 | 1459 | 143 | |
| XM_006249653.2 | XP_006249715.1 | 9 - 10 | 2097 | 143 | ||
| GenBank mRNA | AB121032.1 | - | 8 - 9 | 1459 | 143 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P052815 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 813951 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 619304 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 958605 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 459927 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 602814 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 360672 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 235736 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 762865 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 532419 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 413538 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 755390 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 353243 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 452245 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 492283 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 1961 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 436192 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 723985 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 241878 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 877413 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 120640 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 457996 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 908119 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 905983 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 544595 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 385360 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 570186 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 225890 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 135741 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 735777 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 767259 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 221449 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 832529 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 625125 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 148142 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 869927 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 230965 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 745288 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 456361 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 809272 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 239838 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 436168 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 927271 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 573278 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 161952 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 497421 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 823969 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 119138 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 271187 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 134037 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 41773 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 65492 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 21167 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 104430 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 133830 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 47088 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 6722 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 172543 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 112127 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 59836 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P052815 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P052815 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P052815 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P13646 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 40221 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 76440 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 83208 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 56448 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 86809 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 61793 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 103271 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 76874 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10767017 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10767018 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10767020 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10767022 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10767024 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10767027 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10767025 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10767023 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10767026 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10767019 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10767021 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10767016 | - | - |
| Affymetrix Rat Clariom S Assay | PSR1300008195.rn.1 | TC1300001235.rn.1 | RTA TC |
| Affymetrix Rat Gene 1.0 ST Array | 10767016 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21314109 | - | - |
| ABI Rat Genome Survey Microarray | 21993582 | - | - |
| ABI Rat Genome Survey Microarray | 22169716 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1376619 | - | - |

Cdh19
Genomic Map