Assay Details
| Gene Symbol: Ttc13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001136162.1 | NP_001129634.1 | 12 - 13 | 1538 | 66 | |
| XM_006255802.2 | XP_006255864.1 | 14 - 15 | 1706 | 66 | ||
| XM_006255803.2 | XP_006255865.1 | 14 - 15 | 1706 | 66 | ||
| XM_006255805.2 | XP_006255867.1 | 12 - 13 | 1547 | 66 | ||
| GenBank mRNA | AY724473.1 | - | 2 - 3 | 175 | 66 | |
| BC105871.1 | - | 4 - 5 | 523 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ttc13
Genomic Map