Assay Details
Gene Symbol: Galnt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106196.1 | NP_001099666.1 | 13 - 14 | 1348 | 89 | |
XM_008772605.1 | XP_008770827.1 | 15 - 16 | 5305 | 89 | ||
GenBank mRNA | BC168926.1 | - | 1559 | 89 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P108001 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 361294 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 346025 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 542165 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 348689 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 610125 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 943945 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 119383 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 71023 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 909547 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 610548 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 573264 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 218277 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 211050 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 31735 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 501523 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 139700 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 197470 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 15922 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 436355 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 969756 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 468643 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 823167 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 639741 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 461659 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 212398 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 640504 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 427096 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 481502 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 301182 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 370342 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 85209 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 774869 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 471100 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 69885 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 75302 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 881 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 136006 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 168261 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 129384 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 16304 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 179385 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P108001 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P108001 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P108001 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P471141 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 32845 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 50347 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10808787 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10808783 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10808776 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10808786 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10808775 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10808780 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10808784 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10808789 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10808779 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10808777 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10808778 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10808782 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10808785 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10808788 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10808781 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10808774 | - | - |
Rat Genome 230 2.0 Array | 1375913_at | - | - |
Rat Genome 230 2.0 Array | 1385402_at | - | - |
Rat Genome 230 2.0 Array | 1386440_PM_a_at | - | - |
Rat Genome 230 2.0 Array | 1390775_PM_at | - | - |
Rat Genome 230 2.0 Array | 1375214_PM_at | - | - |
Rat Genome 230 2.0 Array | 1375912_PM_a_at | - | - |
Rat Genome 230 2.0 Array | 1386441_PM_at | - | - |
Rat Genome 230 2.0 Array | 1375913_PM_at | - | - |
Rat Genome 230 2.0 Array | 1390775_at | - | - |
Rat Genome 230 2.0 Array | 1375214_at | - | - |
Rat Genome 230 2.0 Array | 1385402_PM_at | - | - |
Rat Genome 230 2.0 Array | 1386441_at | - | - |
Rat Genome 230 2.0 Array | 1375912_a_at | - | - |
Rat Genome 230 2.0 Array | 1386440_a_at | - | - |
Affymetrix HT Rat Focus Array | 1375913_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1375912_PM_a_at | - | - |
Affymetrix HT Rat Focus Array | 1385402_at | - | - |
Affymetrix HT Rat Focus Array | 1375913_at | - | - |
Affymetrix HT Rat Focus Array | 1375214_at | - | - |
Affymetrix HT Rat Focus Array | 1386441_at | - | - |
Affymetrix HT Rat Focus Array | 1375912_a_at | - | - |
Affymetrix HT Rat Focus Array | 1375214_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1386441_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1385402_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1900006792.rn.1 | TC1900000834.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1900006794.rn.1 | TC1900000834.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1900006791.rn.1 | TC1900000834.rn.1 | RTA TC |
Rat Expression Set 230 | 1385402_at | - | - |
Rat Expression Set 230 | 1375913_PM_at | - | - |
Rat Expression Set 230 | 1385402_PM_at | - | - |
Rat Expression Set 230 | 1375912_PM_a_at | - | - |
Rat Expression Set 230 | 1375913_at | - | - |
Rat Expression Set 230 | 1375214_at | - | - |
Rat Expression Set 230 | 1375214_PM_at | - | - |
Rat Expression Set 230 | 1375912_a_at | - | - |
Rat Expression Set 230 | 1386441_at | - | - |
Rat Expression Set 230 | 1390775_PM_at | - | - |
Rat Expression Set 230 | 1386441_PM_at | - | - |
Rat Expression Set 230 | 1390775_at | - | - |
Rat Expression Set 230 | 1386440_a_at | - | - |
Rat Expression Set 230 | 1386440_PM_a_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10808774 | - | - |
Rat Genome U34 Set | rc_AI011863_at | - | - |
Rat Genome U34 Set | rc_AA963297_at | - | - |
Rat Genome U34 Set | rc_AA997425_at | - | - |
Rat Genome U34 Set | rc_AA859402_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21347371 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1355124 | - | - |
