Assay Details
| Gene Symbol: Capn9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271140.1 | NP_001258069.1 | 16 - 17 | 1822 | 63 | |
| GenBank mRNA | U89514.1 | - | 16 - 17 | 1743 | 63 | |
More Information
Updated Mapping Information
Set Membership: |
|

Capn9
Genomic Map