Assay Details
| Gene Symbol: Pask | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009362.1 | NP_001009362.1 | 15 - 16 | 3970 | 74 | |
| GenBank mRNA | BC087047.1 | - | 15 - 16 | 3970 | 7112450 | 74 |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
protein phosphorylation
signal transduction
regulation of respiratory gaseous exchange
negative regulation of glycogen biosynthetic process
protein autophosphorylation
regulation of glucagon secretion
regulation of energy homeostasis
protein serine/threonine kinase activity
signal transducer activity
ATP binding
phosphatidylinositol binding
protein phosphorylation
signal transduction
regulation of respiratory gaseous exchange
negative regulation of glycogen biosynthetic process
protein autophosphorylation
regulation of glucagon secretion
regulation of energy homeostasis
protein serine/threonine kinase activity
signal transducer activity
ATP binding
phosphatidylinositol binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_43_P18802 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 887699 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 961023 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 696643 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 33823 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 188276 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 774161 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 672645 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 445794 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 894364 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 690252 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 655023 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 417167 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 657627 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 914468 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 154613 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 13800 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 186310 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 660686 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 189068 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 686420 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 293589 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 927553 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 259033 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 67756 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 81270 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 40252 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 54201 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P18802 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P18802 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_43_P18802 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P18802 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 80192 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 91977 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 80031 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 91216 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 73874 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10930012 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10930013 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10929998 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10929999 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10930004 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10929997 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10930010 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10930009 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10930006 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10930008 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10930001 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10930011 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10930007 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10930002 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10929996 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10930014 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10930005 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10930003 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10929995 | - | - |
| Rat Genome 230 2.0 Array | 1392008_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1392008_at | - | - |
| Affymetrix HT Rat Focus Array | 1392008_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1392008_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0900020314.rn.1 | TC0900002478.rn.1 | RTA TC |
| Rat Expression Set 230 | 1392008_at | - | - |
| Rat Expression Set 230 | 1392008_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10929995 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21817646 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1363607 | - | - |
| RatRef-12 Expression BeadChip | ILMN_50061 | - | - |

Pask
Genomic Map