Assay Details
| Gene Symbol: Nudt12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109010.1 | NP_001102480.1 | 4 - 5 | 1076 | 65 | |
| XM_006245588.2 | XP_006245650.1 | 4 - 5 | 1081 | 65 | ||
| XM_006245589.2 | XP_006245651.1 | 4 - 5 | 1085 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nudt12
Genomic Map