Assay Details
Gene Symbol: Wash | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127390.1 | NP_001120862.1 | 1139 | 116 | ||
GenBank mRNA | BC088270.1 | - | 1 - 2 | 181 | 116 | |
BC101888.1 | - | 4 - 5 | 495 | 116 | ||
BC166761.1 | - | 1139 | 7930951 | 116 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
