Assay Details
| Gene Symbol: U2af1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044291.1 | NP_001037756.1 | 3 - 4 | 251 | 78 | |
| GenBank mRNA | BC105905.1 | - | 3 - 4 | 251 | 7374686 | 78 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

U2af1
Genomic Map