Assay Details
Gene Symbol: Kifc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005878.1 | NP_001005878.1 | 7 - 8 | 1654 | 62 | |
GenBank mRNA | AF035951.1 | - | 1 - 2 | 328 | 62 | |
BC083827.1 | - | 7 - 8 | 1654 | 7112250 | 62 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
