Assay Details
| Gene Symbol: RGD1303003 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004225.1 | NP_001004225.1 | 1 - 2 | 148 | 91 | |
| GenBank mRNA | BC079380.1 | - | 1 - 2 | 148 | 7110981 | 91 |
| FQ217777.1 | - | 1 - 2 | 164 | 91 | ||
| FQ231770.1 | - | 1 - 2 | 145 | 91 | ||
More Information
Updated Mapping Information
Set Membership: |
|

RGD1303003
Genomic Map