Assay Details
Gene Symbol: RGD1303003 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004225.1 | NP_001004225.1 | 3 - 4 | 311 | 87 | |
GenBank mRNA | BC079380.1 | - | 3 - 4 | 311 | 7110981 | 87 |
FQ213331.1 | - | 3 - 4 | 321 | 87 | ||
FQ217777.1 | - | 3 - 4 | 327 | 87 | ||
FQ231770.1 | - | 3 - 4 | 308 | 87 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
