Assay Details
Gene Symbol: RGD1303003 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004225.1 | NP_001004225.1 | 4 - 5 | 432 | 92 | |
GenBank mRNA | BC079380.1 | - | 4 - 5 | 432 | 7110981 | 92 |
FQ213331.1 | - | 4 - 5 | 442 | 92 | ||
FQ217777.1 | - | 4 - 5 | 448 | 92 | ||
FQ231770.1 | - | 4 - 5 | 429 | 92 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
