Assay Details
| Gene Symbol: Slc37a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011944.1 | NP_001011944.1 | 18 - 19 | 1897 | 91 | |
| XM_008772807.1 | XP_008771029.1 | 16 - 17 | 1630 | 91 | ||
| GenBank mRNA | BC081990.1 | - | 18 - 19 | 1897 | 7129340 | 91 |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P096482 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 348867 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 770339 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 197585 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 753022 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 647871 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 303380 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 295639 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 927719 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 477804 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 265644 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 44755 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 62602 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 936515 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 710361 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 888527 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 184607 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 323393 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 806605 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 450766 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 568451 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 613024 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 119119 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 151400 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 219284 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 114025 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 427486 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 258431 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 827244 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 804123 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 848267 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 762231 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 734253 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 805284 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 904379 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 888915 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 816468 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 670281 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 683640 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 834892 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 548452 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 761033 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 786417 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 840846 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 177416 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 63811 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 34703 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 77111 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 99865 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 101118 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P096482 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P096482 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P096482 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P20478 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 38076 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 544 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 27053 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10829121 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10829120 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10829131 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10829128 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10829118 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10829125 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10829135 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10829136 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10829119 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10829133 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10829134 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10829124 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10829123 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10829129 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10829117 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10829127 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10829122 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10829126 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10829132 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10829130 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10829116 | - | - |
| Rat Genome 230 2.0 Array | 1382076_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1382076_at | - | - |
| Affymetrix HT Rat Focus Array | 1382076_at | - | - |
| Affymetrix HT Rat Focus Array | 1382076_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR2000003398.rn.1 | TC2000000358.rn.1 | RTA TC |
| Rat Expression Set 230 | 1382076_at | - | - |
| Rat Expression Set 230 | 1382076_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10829116 | - | - |
| Rat Genome U34 Set | rc_AA799487_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21128602 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1376690 | - | - |

Slc37a1
Genomic Map