Assay Details
| Gene Symbol: Snrpd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003751948.3 | XP_003751996.1 | 1 - 2 | 128 | 75 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_003751948.3 transcripts.
Set Membership: |
|

Snrpd3
Genomic Map