Assay Details
| Gene Symbol: RGD1564149 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079708.1 | NP_001073176.1 | 1 - 2 | 244 | 64 | |
| GenBank mRNA | BC128753.1 | - | 1 - 2 | 244 | 7100076 | 64 |
More Information
Updated Mapping Information
Set Membership: |
|

RGD1564149
Genomic Map