Assay Details
Gene Symbol: RGD1564450 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106377.1 | NP_001099847.1 | 2 - 3 | 468 | 102 | |
XM_006256171.2 | XP_006256233.1 | 2 - 3 | 286 | 102 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001106377.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
